Tisk…

4,4-Dimethoxybutan-2-ol 95%

Dodavatel: Apollo Scientific

OR924280-1G
APOSOR924280-1GEA 16340 CZK
APOSOR924280-1G
4,4-Dimethoxybutan-2-ol 95%
4,4-Dimethoxybutan-2-ol
Vzorec: C₆H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD00046655
CAS číslo: 39562-58-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR