Tisk…

2-Methoxy-6-(methylamino)pyridine 95%

Dodavatel: Apollo Scientific

OR923913-25G
APOSOR923913-25GEA 1820 CZK
APOSOR923913-25G
2-Methoxy-6-(methylamino)pyridine 95%
2-Methoxy-6-(methylamino)pyridine
Vzorec: C₇H₁₀N₂O
MW: 138.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191557
CAS číslo: 88569-83-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR