Tisk…

1-Methylpyrrolidine-2-thione 96%

Dodavatel: Apollo Scientific

OR923785-5G
APOSOR923785-5GEA 2070 CZK
APOSOR923785-5G
1-Methylpyrrolidine-2-thione 96%
1-Methylpyrrolidine-2-thione
Vzorec: C₅H₉NS
MW: 115,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00966332
CAS číslo: 10441-57-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR