Tisk…

2-(Methylthio)thiazole 98%

Dodavatel: Apollo Scientific

OR923771-100G
APOSOR923771-100GEA 4730 CZK
APOSOR923771-100G
2-(Methylthio)thiazole 98%
2-(Methylthio)thiazole
Vzorec: C₄H₅NS₂
Storage Temperature: Ambient
MDL Number: MFCD00858981
CAS číslo: 5053-24-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR