Tisk…

Dimethyl succinate 95%

Dodavatel: Apollo Scientific

OR923432-100G
APOSOR923432-100GEA 744 CZK
APOSOR923432-100G
Dimethyl succinate 95%
Dimethyl succinate
Vzorec: C₆H₁₀O₄
Storage Temperature: Ambient
MDL Number: MFCD00008466
CAS číslo: 106-65-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR