Tisk…

1H-Pyrrolo[2,3-b]pyridin-3-ylmethanol 95%

Dodavatel: Apollo Scientific

OR923206-1G OR923206-250MG
APOSOR923206-1GEA 3110 CZK
APOSOR923206-1G APOSOR923206-250MG
1H-Pyrrolo[2,3-b]pyridin-3-ylmethanol 95%
1H-Pyrrolo[2,3-b]pyridin-3-ylmethanol
Vzorec: C₈H₈N₂O
Storage Temperature: Ambient
MDL Number: MFCD11054203
CAS číslo: 1065100-83-5

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR