Tisk…

(1-Ethylazetidin-3-yl)methanamine

Dodavatel: Apollo Scientific

OR922991-1G
APOSOR922991-1GEA 26290 CZK
APOSOR922991-1G
(1-Ethylazetidin-3-yl)methanamine
(1-Ethylazetidin-3-yl)methanamine
Vzorec: C₆H₁₄N₂
Storage Temperature: Ambient
MDL Number: MFCD21661081
CAS číslo: 1412978-07-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR