Tisk…

4-Ethylaniline 98%

Dodavatel: Apollo Scientific

OR922870-500G
APOSOR922870-500GEA 3710 CZK
APOSOR922870-500G
4-Ethylaniline 98%
4-Ethylaniline
Vzorec: C₈H₁₁N
Storage Temperature: Ambient
MDL Number: MFCD00007921
CAS číslo: 589-16-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR