Tisk…

3-Iodopropionic acid 95%

Dodavatel: Apollo Scientific

OR922011-25G
APOSOR922011-25GEA 2940 CZK
APOSOR922011-25G
3-Iodopropionic acid 95%
3-Iodopropionic acid
Vzorec: C₃H₅IO₂
Storage Temperature: Ambient
MDL Number: MFCD00002765
CAS číslo: 141-76-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR