Tisk…

2-(4-Methylpiperazin-1-ylmethyl)phenylamine 97%

Dodavatel: Apollo Scientific
Nebezpečí

OR9219-1G OR9219-250MG
APOSOR9219-1GEA 10620 CZK
APOSOR9219-1G APOSOR9219-250MG
2-(4-Methylpiperazin-1-ylmethyl)phenylamine 97%
2-(4-Methylpiperazin-1-ylmethyl)phenylamine
Vzorec: C₁₂H₁₉N₃
Bod tání: 82…84 °C
Storage Temperature: Ambient
MDL Number: MFCD06804544
CAS číslo: 19577-84-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR