Tisk…

Allyl chloroacetate 95%

Dodavatel: Apollo Scientific

OR921089-25G
APOSOR921089-25GEA 1580 CZK
APOSOR921089-25G
Allyl chloroacetate 95%
Allyl chloroacetate
Vzorec: C₅H₇ClO₂
Storage Temperature: Ambient
MDL Number: MFCD00058936
CAS číslo: 2916-14-5

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR