Tisk…

2-Morpholin-4-ylmethylbenzylamine 95+%

Dodavatel: Apollo Scientific

OR9210-10G OR9210-1G OR9210-5G
APOSOR9210-10GEA 23380 CZK
APOSOR9210-10G APOSOR9210-1G APOSOR9210-5G
2-Morpholin-4-ylmethylbenzylamine 95+%
2-Morpholin-4-ylmethylbenzylamine
Vzorec: C₁₂H₁₈N₂O
Storage Temperature: Ambient
MDL Number: MFCD06655814
CAS číslo: 91271-82-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR