Tisk…

sec-Butyl crotonate 95%

Dodavatel: Apollo Scientific

OR920185-100G OR920185-25G OR920185-5G
APOSOR920185-100GEA 16340 CZK
APOSOR920185-100G APOSOR920185-25G APOSOR920185-5G
sec-Butyl crotonate 95%
sec-Butyl crotonate
Vzorec: C₈H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD00059054
CAS číslo: 10371-45-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR