Tisk…

7-Methyl-1,5-benzodioxepan-3-one 95%

Dodavatel: Apollo Scientific

Synonyms: 7-Methyl-3,4-dihydro-2H-1,5-benzodioxepin-3-one, Watermelon ketone, 7-Methyl-1,5-benzodioxepin-3-one

OR916678-25G
APOSOR916678-25GEA 4630 CZK
APOSOR916678-25G
7-Methyl-1,5-benzodioxepan-3-one 95%
7-Methyl-1,5-benzodioxepan-3-one
Vzorec: C₁₀H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD07371373
CAS číslo: 28940-11-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR