Tisk…

Indoline-5-carboxylic acid hydrochloride 95%

Dodavatel: Apollo Scientific

OR916518-1G OR916518-250MG
APOSOR916518-1GEA 22950 CZK
APOSOR916518-1G APOSOR916518-250MG
Indoline-5-carboxylic acid hydrochloride 95%
Indoline-5-carboxylic acid hydrochloride
Vzorec: C₉H₉NO₂·ClH
Storage Temperature: Ambient
MDL Number: MFCD09971545
CAS číslo: 1171876-52-0

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR