Tisk…

Boc-d-homoserine

Dodavatel: Apollo Scientific
Varování

OR916500-1G OR916500-25G OR916500-5G
APOSOR916500-1GEA 2110 CZK
APOSOR916500-1G APOSOR916500-25G APOSOR916500-5G
Boc-d-homoserine
Boc-d-homoserine
Vzorec: C₉H₁₇NO₅
Storage Temperature: Ambient
MDL Number: MFCD12545888
CAS číslo: 745011-75-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR