Tisk…

(S)-3-Amino-3-(3-methylphenyl)propionic acid 97%

Dodavatel: Apollo Scientific

OR913925-1G
APOSOR913925-1GEA 6620 CZK
APOSOR913925-1G
(S)-3-Amino-3-(3-methylphenyl)propionic acid 97%
(S)-3-Amino-3-(3-methylphenyl)propionic acid
Vzorec: C₁₀H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD04113692
CAS číslo: 701907-44-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR