Tisk…

3-(6-Aminopyridin-3-yl)acrylic acid ethyl ester 95%

Dodavatel: Apollo Scientific

Synonyms: Ethyl 3-(6-aminopyridin-3-yl)acrylate

OR912986-1G
APOSOR912986-1GEA 3870 CZK
APOSOR912986-1G
3-(6-Aminopyridin-3-yl)acrylic acid ethyl ester 95%
3-(6-Aminopyridin-3-yl)acrylic acid ethyl ester
Vzorec: C₁₀H₁₂N₂O₂
MW: 192.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04039876
CAS číslo: 227963-57-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR