Tisk…

2-Chloro-4-nitropyridine-N-oxide 98%

Dodavatel: Apollo Scientific

OR912604-100G OR912604-25G
APOSOR912604-100GEA 13660 CZK
APOSOR912604-100G APOSOR912604-25G
2-Chloro-4-nitropyridine-N-oxide 98%
2-Chloro-4-nitropyridine-N-oxide
Vzorec: C₅H₃ClN₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00955617
CAS číslo: 14432-16-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR