Tisk…

1-(4-Methoxyphenyl)butane-1,3-dione 95%

Dodavatel: Apollo Scientific

OR912527-1G
APOSOR912527-1GEA 2650 CZK
APOSOR912527-1G
1-(4-Methoxyphenyl)butane-1,3-dione 95%
1-(4-Methoxyphenyl)butane-1,3-dione
Vzorec: C₁₁H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00463138
CAS číslo: 4023-80-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR