Tisk…

(R)-2-Amino-3-(4-cyanophenyl)propanoic acid 97%

Dodavatel: Apollo Scientific

Synonyms: 4-Cyano-D-phenylalanine, H-4-Cyano-D-Phe-OH, H-D-Phe(4-CN)-OH

OR912500-5G
APOSOR912500-5GEA 2110 CZK
APOSOR912500-5G
(R)-2-Amino-3-(4-cyanophenyl)propanoic acid 97%
(R)-2-Amino-3-(4-cyanophenyl)propanoic acid
Vzorec: C₁₀H₁₀N₂O₂
MW: 190.202 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00270364
CAS číslo: 263396-44-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR