Tisk…

2,3-Dimethoxy-5-methylhydroquinone 98%

Dodavatel: Apollo Scientific

Synonyms: 2,3-Dimethoxy-5-methylbenzene-1,4-diol

OR911920-1G
APOSOR911920-1GEA 4770 CZK
APOSOR911920-1G
2,3-Dimethoxy-5-methylhydroquinone 98%
2,3-Dimethoxy-5-methylhydroquinone
Vzorec: C₉H₁₂O₄
MW: 184.189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00017161
CAS číslo: 3066-90-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR