Tisk…

2-Etylpiperidin 95%

Dodavatel: Apollo Scientific

OR911632-25G
APOSOR911632-25GEA 4860 CZK
APOSOR911632-25G
2-Etylpiperidin 95%
2-Etylpiperidin
Vzorec: C₇H₁₅N
MW: 113.2
Storage Temperature: Ambient
MDL Number: MFCD00005988
CAS číslo: 1484-80-6

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR