Tisk…

tert-Butyl-5-norbornene-2-carboxylate (endo and exo mixture) 95%

Dodavatel: Apollo Scientific
Varování

OR911625-25G OR911625-5G
APOSOR911625-25GEA 1080 CZK
APOSOR911625-25G APOSOR911625-5G
tert-Butyl-5-norbornene-2-carboxylate (endo and exo mixture) 95%
tert-Butyl-5-norbornene-2-carboxylate (endo and exo mixture)
Vzorec: C₁₂H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD18083704
CAS číslo: 154970-45-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR