Tisk…

1-benzylpyrrole-2-carbaldehyde 95%

Dodavatel: Apollo Scientific

OR910391-1G OR910391-250MG
APOSOR910391-1GEA 24600 CZK
APOSOR910391-1G APOSOR910391-250MG
1-benzylpyrrole-2-carbaldehyde 95%
1-benzylpyrrole-2-carbaldehyde
Vzorec: C₁₂H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD09674969
CAS číslo: 18159-24-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR