Tisk…

6-Chloro-4-methylquinolin-2(1H)-one 98%

Dodavatel: Apollo Scientific

OR908968-1G OR908968-5G
APOSOR908968-1GEA 12830 CZK
APOSOR908968-1G APOSOR908968-5G
6-Chloro-4-methylquinolin-2(1H)-one 98%
6-Chloro-4-methylquinolin-2(1H)-one
Vzorec: C₁₀H₈ClNO
Storage Temperature: Ambient
MDL Number: MFCD02269197
CAS číslo: 2585-04-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR