Tisk…

2-Isopropoxy-5-methylpyridine-3-boronic acid 98%

Dodavatel: Apollo Scientific

OR908561-1G OR908561-5G
APOSOR908561-1GEA 6220 CZK
APOSOR908561-1G APOSOR908561-5G
2-Isopropoxy-5-methylpyridine-3-boronic acid 98%
2-Isopropoxy-5-methylpyridine-3-boronic acid
Vzorec: C₉H₁₄BNO₃
Storage Temperature: Ambient
MDL Number: MFCD19237189
CAS číslo: 2096337-70-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR