Tisk…

2-Cyano-2-methylpropanoic acid 98%

Dodavatel: Apollo Scientific

OR908203-1G OR908203-5G
APOSOR908203-1GEA 1820 CZK
APOSOR908203-1G APOSOR908203-5G
2-Cyano-2-methylpropanoic acid 98%
2-Cyano-2-methylpropanoic acid
Vzorec: C₅H₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD11100210
CAS číslo: 22426-30-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR