Tisk…

1-Boc-4-cyano-4-methylpiperidine 97%

Dodavatel: Apollo Scientific

OR907558-1G OR907558-250MG OR907558-500MG
APOSOR907558-1GEA 16370 CZK
APOSOR907558-1G APOSOR907558-250MG APOSOR907558-500MG
1-Boc-4-cyano-4-methylpiperidine 97%
1-Boc-4-cyano-4-methylpiperidine
Vzorec: C₁₂H₂₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD11849789
CAS číslo: 530115-96-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR