Tisk…

4-(Morpholinomethyl)phenylboronic acid hydrochloride ≥96%

Dodavatel: Apollo Scientific

OR907494-1G OR907494-250MG
APOSOR907494-1GEA 2860 CZK
APOSOR907494-1G APOSOR907494-250MG
4-(Morpholinomethyl)phenylboronic acid hydrochloride ≥96%
4-(Morpholinomethyl)phenylboronic acid hydrochloride
Vzorec: C₁₁H₁₆BNO₃·ClH
Storage Temperature: Refrigerator
MDL Number: MFCD16661158
CAS číslo: 1335234-36-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR