Tisk…

4-Bromo-5-chloroanthranilic acid 98%

Dodavatel: Apollo Scientific

Synonyms: 2-Amino-4-bromo-5-chlorobenzoic acid

OR907297-1G OR907297-25G OR907297-5G
APOSOR907297-1GEA 622 CZK
APOSOR907297-1G APOSOR907297-25G APOSOR907297-5G
4-Bromo-5-chloroanthranilic acid 98%
4-Bromo-5-chloroanthranilic acid
Vzorec: C₇H₅BrClNO₂
MW: 250.4800 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10000554
CAS číslo: 150812-32-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR