Tisk…

4-sec-Butylaniline 98%

Dodavatel: Apollo Scientific

Synonyms: 4-(sec-Butyl)aniline

OR906980-5G
APOSOR906980-5GEA 1870 CZK
APOSOR906980-5G
4-sec-Butylaniline 98%
4-sec-Butylaniline
Vzorec: C₁₀H₁₅N
MW: 149.24
Bod varu: 238
Storage Temperature: Ambient
MDL Number: MFCD00010223
CAS číslo: 30273-11-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR