Tisk…

N-(tert-Butoxycarbonyl)propargylamine 98%

Dodavatel: Apollo Scientific

OR906557-100G
APOSOR906557-100GEA 4370 CZK
APOSOR906557-100G
N-(tert-Butoxycarbonyl)propargylamine 98%
N-(tert-Butoxycarbonyl)propargylamine
Vzorec: C₈H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD07367245
CAS číslo: 92136-39-5

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR