Tisk…

1,3-Dimethyl-1H-pyrazol-4-carbaldehyde 98%

Dodavatel: Apollo Scientific

OR904769-1G OR904769-25G OR904769-5G
APOSOR904769-1GEA 3540 CZK
APOSOR904769-1G APOSOR904769-25G APOSOR904769-5G
1,3-Dimethyl-1H-pyrazol-4-carbaldehyde 98%
1,3-Dimethyl-1H-pyrazol-4-carbaldehyde
Vzorec: C₆H₈N₂O
Storage Temperature: Ambient
MDL Number: MFCD00159635
CAS číslo: 25016-12-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR