Tisk…

4-(2-Formyl-1-pyrrolyl)benzonitrile 95%

Dodavatel: Apollo Scientific

OR904656-1G OR904656-250MG
APOSOR904656-1GEA 9330 CZK
APOSOR904656-1G APOSOR904656-250MG
4-(2-Formyl-1-pyrrolyl)benzonitrile 95%
4-(2-Formyl-1-pyrrolyl)benzonitrile
Vzorec: C₁₂H₈N₂O
Storage Temperature: Ambient
MDL Number: MFCD02664880
CAS číslo: 169036-66-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR