Tisk…

Tetrabutylammonium tribromide 97%

Dodavatel: Apollo Scientific
Nebezpečí

OR902166-100G OR902166-500G
APOSOR902166-100GEA 3670 CZK
APOSOR902166-100G APOSOR902166-500G
Tetrabutylammonium tribromide 97%
Tetrabutylammonium tribromide
Vzorec: C₁₆H₃₆Br₃N
Bod tání: 71…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00012110
CAS číslo: 38932-80-8
UN: 1759
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR