Tisk…

5-Bromo-1-methylpyridin-2(1H)-one 98%

Dodavatel: Apollo Scientific

Synonyms: 5-Bromo-1-methyl-2(1H)-pyridinone

OR901973-5G
APOSOR901973-5GEA 1740 CZK
APOSOR901973-5G
5-Bromo-1-methylpyridin-2(1H)-one 98%
5-Bromo-1-methylpyridin-2(1H)-one
Vzorec: C₆H₆BrNO
MW: 188.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09972188
CAS číslo: 81971-39-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR