Tisk…

N-tert-Butylisonicotinamide 98%

Dodavatel: Apollo Scientific

OR901667-1G OR901667-25G OR901667-5G
APOSOR901667-1GEA 1450 CZK
APOSOR901667-1G APOSOR901667-25G APOSOR901667-5G
N-tert-Butylisonicotinamide 98%
N-tert-Butylisonicotinamide
Vzorec: C₁₀H₁₄N₂O
Storage Temperature: Ambient
MDL Number: MFCD00466497
CAS číslo: 65321-30-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR