Tisk…

4-Bromothiophene-2-methanol 98%

Dodavatel: Apollo Scientific

OR900842-25G
APOSOR900842-25GEA 3230 CZK
APOSOR900842-25G
4-Bromothiophene-2-methanol 98%
4-Bromothiophene-2-methanol
Vzorec: C₅H₅BrOS
Storage Temperature: Ambient
MDL Number: MFCD04115392
CAS číslo: 79757-77-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR