Tisk…

Propoxybenzene 98%

Dodavatel: Apollo Scientific

OR900706-1G OR900706-5G
APOSOR900706-1GEA 953 CZK
APOSOR900706-1G APOSOR900706-5G
Propoxybenzene 98%
Propoxybenzene
Vzorec: C₉H₁₂O
Storage Temperature: Ambient
MDL Number: MFCD00039935
CAS číslo: 622-85-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR