Tisk…

5-Chloro-4-methyl-2-nitrophenol 98%

Dodavatel: Apollo Scientific

OR900151-1G
APOSOR900151-1GEA 10350 CZK
APOSOR900151-1G
5-Chloro-4-methyl-2-nitrophenol 98%
5-Chloro-4-methyl-2-nitrophenol
Vzorec: C₇H₆ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD19374209
CAS číslo: 100278-74-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR