Tisk…

(R)-(-)-Ethyl-3-piperidinecarboxylate

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Ethyl (R)-(-)-3-Piperidinecarboxylate, (R)-Ethyl piperidine-3-carboxylate

OR8969-100G OR8969-25G OR8969-5G
APOSOR8969-100GEA 5260 CZK
APOSOR8969-100G APOSOR8969-25G APOSOR8969-5G
(R)-(-)-Ethyl-3-piperidinecarboxylate
(R)-(-)-Ethyl-3-piperidinecarboxylate
Vzorec: C₈H₁₅NO₂
MW: 157.2102 g/mol
Hustota: 1.092 g/cm³ (25 °C)
Bod vzplanutí: 104 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03093637
CAS číslo: 25137-01-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR