Tisk…

11-Azido-3,6,9-trioxaundecan-1-amine 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-Amino-11-azido-3,6,9-trioxaundecane, Amino-PEG3-Azide, 11-Amino-3,6,9-trioxaundecanyl Azide, Azido-PEG3-Amine

OR8503T-1G OR8503T-5G
APOSOR8503T-1GEA 4570 CZK
APOSOR8503T-1G APOSOR8503T-5G
11-Azido-3,6,9-trioxaundecan-1-amine 97%
11-Azido-3,6,9-trioxaundecan-1-amine

Heterobifunctional PRG derivative. Contains a free aminethat can be conjugated to biological molecules directly by an amide linkage (or via the corresponding isothiocyanate).

Vzorec: C₈H₁₈N₄O₃
MW: 218.26 g/mol
Bod varu: 198…202 °C
Hustota: 1.1 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00269874
CAS číslo: 134179-38-7
UN: 2735
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR