Tisk…

2-Phenylpyrimidine

Dodavatel: Apollo Scientific
Varování

OR8422-1G
APOSOR8422-1GEA 7340 CZK
APOSOR8422-1G
2-Phenylpyrimidine
2-Phenylpyrimidine
Vzorec: C₁₀H₈N₂
MW: 156.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04038761
CAS číslo: 7431-45-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR