Tisk…

2-Methyl-5-(piperidin-1-ylsulphonyl)-3-furoic acid 95%

Dodavatel: Apollo Scientific
Nebezpečí

OR8133-1G OR8133-250MG
APOSOR8133-1GEA 3970 CZK
APOSOR8133-1G APOSOR8133-250MG
2-Methyl-5-(piperidin-1-ylsulphonyl)-3-furoic acid 95%
2-Methyl-5-(piperidin-1-ylsulphonyl)-3-furoic acid
Vzorec: C₁₁H₁₅NO₅S
Bod tání: 188…189 °C
Storage Temperature: Ambient
MDL Number: MFCD02180759
CAS číslo: 306936-38-9
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR