Tisk…

3,6-Dichloropicolinic acid

Dodavatel: Apollo Scientific
Nebezpečí

OR8123-100G OR8123-25G OR8123-500G
APOSOR8123-100GEA 1660 CZK
APOSOR8123-100G APOSOR8123-25G APOSOR8123-500G
3,6-Dichloropicolinic acid
3,6-Dichloropicolinic acid
Vzorec: C₆H₃Cl₂NO₂
Bod vzplanutí: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00078655
CAS číslo: 1702-17-6
EINECS: 216-935-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR