Tisk…

Boc-py-oh 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-((tert-Butoxycarbonyl)amino)-1-methyl-1H-pyrrole-2-carboxylic acid, 4-(Boc-amino)-1-methyl-1H-pyrrole-2-carboxylic acid

OR8042-1G OR8042-250MG
APOSOR8042-1GEA 2280 CZK
APOSOR8042-1G APOSOR8042-250MG
Boc-py-oh 97%
Boc-py-oh
Vzorec: C₁₁H₁₆N₂O₄
MW: 240.259 g/mol
Bod tání: 156…160 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD02094544
CAS číslo: 77716-11-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR