Tisk…

1-Acetyl-2,3-dihydro-N-[1-(2-phenylethyl)piperidin-4-yl]-1H-indole-5-amine 95+%

Dodavatel: Apollo Scientific

OR7925-1G OR7925-5G OR7925-25G
APOSOR7925-1GEA 5260 CZK
APOSOR7925-1G APOSOR7925-5G APOSOR7925-25G
1-Acetyl-2,3-dihydro-N-[1-(2-phenylethyl)piperidin-4-yl]-1H-indole-5-amine 95+%
1-Acetyl-2,3-dihydro-N-[1-(2-phenylethyl)piperidin-4-yl]-1H-indole-5-amine
Vzorec: C₂₃H₂₉N₃O
MW: 363.51 g/mol
Bod tání: 156…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00214533
CAS číslo: 394653-85-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR