Tisk…

2-Bromophenylhydrazinium chloride 98%

Dodavatel: Apollo Scientific
Nebezpečí

OR7758-100G OR7758-250G OR7758-25G
APOSOR7758-100GEA 1910 CZK
APOSOR7758-100G APOSOR7758-250G APOSOR7758-25G
2-Bromophenylhydrazinium chloride 98%
2-Bromophenylhydrazinium chloride
Vzorec: C₆H₈BrClN₂
Storage Temperature: Ambient
MDL Number: MFCD00012926
CAS číslo: 50709-33-6
EINECS: 256-728-6
UN: 1759
ADR: 8,II

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR