Tisk…

1-tert-Butoxycarbonyl-4-(cyclopropylamino)piperidine 95+%

Dodavatel: Apollo Scientific
Varování

OR7703-1G OR7703-250MG
APOSOR7703-1GEA 3970 CZK
APOSOR7703-1G APOSOR7703-250MG
1-tert-Butoxycarbonyl-4-(cyclopropylamino)piperidine 95+%
1-tert-Butoxycarbonyl-4-(cyclopropylamino)piperidine
Vzorec: C₁₃H₂₄N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD03426074
CAS číslo: 179557-01-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR